siRNA Details
Gene Symbol: Lgalsl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134730.1 | NP_001128202.1 | 5 | 652 | ||
GenBank mRNA | BC168193.1 | - | 5 | 652 | 7109522 | |
FQ211679.1 | - | 5 | 496 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_173752, AK041053, AK081513, BC019131, AK156350, and others.
Gene Ontology Categories:
