siRNA Details
Gene Symbol: LOC682206 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008758828.1 | XP_008757050.1 | 4 | 487 | ||
XM_008758829.1 | XP_008757051.1 | 3 | 456 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC682206 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008758828.1 | XP_008757050.1 | 4 | 487 | ||
XM_008758829.1 | XP_008757051.1 | 3 | 456 |