siRNA Details
Gene Symbol: LOC682206 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008758828.1 | XP_008757050.1 | 4 | 751 | ||
XM_008758829.1 | XP_008757051.1 | 3 | 720 |

More Information
Other Species Targeted

