siRNA Details
Gene Symbol: Setdb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271175.1 | NP_001258104.1 | 6, 7 | 790 | ||
XM_006232913.2 | XP_006232975.1 | 7, 8 | 1180 | |||
XM_006232914.2 | XP_006232976.1 | 6, 7 | 961 | |||
XM_006232916.2 | XP_006232978.1 | 2, 3 | 157 | |||
XM_006232917.2 | XP_006232979.1 | 2, 3 | 660 |
Gene Symbol: LOC100911180 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003752173.3 | XP_003752221.2 | 2, 3 | 238 | ||
XM_008773705.1 | XP_008771927.1 | 1, 2 | 802 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
