siRNA Details
Gene Symbol: Catsperd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134984.1 | NP_001128456.1 | 3, 4 | 240 | ||
XM_006244339.2 | XP_006244401.1 | 3, 4 | 1569 | |||
XM_006244340.2 | XP_006244402.1 | 3, 4 | 1260 | |||
XM_006244342.2 | XP_006244404.1 | 3, 4 | 1569 | |||
XM_006244343.2 | XP_006244405.1 | 3, 4 | 1569 | |||
XM_006244344.2 | XP_006244406.1 | 3, 4 | 1569 | |||
XM_008766771.1 | XP_008764993.1 | 3, 4 | 1569 | |||
GenBank mRNA | BC169121.1 | - | 3, 4 | 240 | 7133439 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
