siRNA Details
Gene Symbol: LOC681325 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109439.1 | NP_001102909.1 | 2 | 831 | ||
GenBank mRNA | FQ226125.1 | - | 2 | 868 |

More Information
Other Species Targeted

