siRNA Details
Gene Symbol: Acyp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001169145.1 | NP_001162616.1 | 4 | 456 | ||
GenBank mRNA | CB613181.1 | - | 4 | 466 | ||
CO404339.1 | - | 4 | 287 | |||
FQ215015.1 | - | 4 | 288 | |||
FQ224926.1 | - | 4 | 324 |

More Information
Other Species Targeted

Gene Ontology Categories:
