siRNA Details
Gene Symbol: Cluap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014225.1 | NP_001014247.1 | 5 | 799 | ||
XM_006245832.2 | XP_006245894.1 | 5 | 562 | |||
XM_008767512.1 | XP_008765734.1 | 5 | 495 | |||
XM_008767513.1 | XP_008765735.1 | 5 | 857 | |||
XM_008767516.1 | XP_008765738.1 | 4 | 401 | |||
XM_008767517.1 | XP_008765739.1 | 3 | 264 | |||
XM_008767518.1 | XP_008765740.1 | 3 | 321 | |||
GenBank mRNA | BC079022.1 | - | 5 | 799 | 7113868 |
