siRNA Details
Gene Symbol: Polr3g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109468.1 | NP_001102938.1 | 4 | 978 | ||
XM_006231729.2 | XP_006231791.1 | 3 | 320 | |||
GenBank mRNA | FQ209718.1 | - | 4 | 737 | ||
FQ210837.1 | - | 4 | 737 |
Gene Symbol: LOC103691413 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760643.1 | XP_008758865.1 | 1 | 894 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
