siRNA Details
Gene Symbol: Ciart | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008761317.1 | XP_008759539.1 | 4 | 578 | ||
XM_008761318.1 | XP_008759540.1 | 5 | 533 | |||
XM_008761319.1 | XP_008759541.1 | 5 | 533 | |||
XM_008761320.1 | XP_008759542.1 | 4 | 356 |
