siRNA Details
Gene Symbol: Pknox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271278.1 | NP_001258207.1 | 6 | 726 | ||
NM_001271279.1 | NP_001258208.1 | 7 | 858 | |||
GenBank mRNA | FQ221048.1 | - | 7 | 858 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
