siRNA Details
Gene Symbol: Kif28p | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270385.1 | NP_001257314.1 | 23 | 2977 | ||
GenBank mRNA | AB573712.1 | - | 23 | 2977 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
