siRNA Details
Gene Symbol: Txlna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127633.1 | NP_001121105.1 | 3 | 454 | ||
XM_006238972.2 | XP_006239034.1 | 3 | 461 | |||
GenBank mRNA | BC166486.1 | - | 3 | 454 | 7099983 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
