siRNA Details
Gene Symbol: Elovl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109118.1 | NP_001102588.1 | 7, 8 | 886 | ||
XM_006253779.2 | XP_006253841.1 | 7, 8 | 832 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
