siRNA Details
Gene Symbol: Wasl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110365.1 | NP_001103835.1 | 9 | 1583 | ||
GenBank mRNA | D88461.1 | - | 9 | 1289 |

More Information
Other Species Targeted

Gene Ontology Categories:
