siRNA Details
Gene Symbol: Clnk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006251106.2 | XP_006251168.1 | 6, 7 | 563 | ||
XM_006251108.2 | XP_006251170.1 | 6, 7 | 563 | |||
XM_008770228.1 | XP_008768450.1 | 3, 4 | 409 | |||
XM_008770229.1 | XP_008768451.1 | 3, 4 | 409 | |||
XM_008770230.1 | XP_008768452.1 | 3, 4 | 409 |
