siRNA Details
Gene Symbol: LOC103689981 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008758849.1 | XP_008757071.1 | 1327 |
Gene Symbol: Vom2r19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099492.1 | NP_001092962.1 | 4 | 1327 |
