siRNA Details
Gene Symbol: Slbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130988.1 | NP_001124460.1 | 5 | 444 | ||
GenBank mRNA | BC167755.1 | - | 5 | 444 | 8363815 |

More Information
Other Species Targeted

Gene Ontology Categories:
