siRNA Details
Gene Symbol: Dazl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109414.1 | NP_001102884.1 | 9 | 932 | ||
XM_006244477.2 | XP_006244539.1 | 10 | 836 |

More Information
Other Species Targeted

Gene Ontology Categories:
