siRNA Details
Gene Symbol: Cycs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012839.2 | NP_036971.1 | 3 | 282 | ||
GenBank mRNA | BC081849.1 | - | 3 | 282 | 7192255 | |
BC089051.1 | - | 3 | 293 | 7305562 | ||
FQ211631.1 | - | 3 | 274 | |||
FQ212556.1 | - | 3 | 276 | |||
FQ212860.1 | - | 3 | 334 | |||
FQ213464.1 | - | 3 | 274 | |||
FQ214646.1 | - | 3 | 305 | |||
FQ214733.1 | - | 3 | 274 | |||
FQ214738.1 | - | 3 | 272 | |||
FQ214855.1 | - | 3 | 305 | |||
FQ214908.1 | - | 3 | 274 | |||
FQ215052.1 | - | 3 | 274 | |||
FQ215109.1 | - | 3 | 274 | |||
FQ215466.1 | - | 3 | 274 | |||
FQ216163.1 | - | 3 | 273 | |||
FQ216217.1 | - | 3 | 272 | |||
FQ216281.1 | - | 3 | 332 | |||
FQ216426.1 | - | 3 | 274 | |||
FQ216512.1 | - | 3 | 274 | |||
FQ216560.1 | - | 3 | 274 | |||
FQ216674.1 | - | 3 | 273 | |||
FQ216679.1 | - | 3 | 274 | |||
FQ217283.1 | - | 3 | 274 | |||
FQ217634.1 | - | 3 | 274 | |||
FQ218428.1 | - | 3 | 274 | |||
FQ218668.1 | - | 3 | 275 | |||
FQ220222.1 | - | 3 | 305 | |||
FQ220855.1 | - | 3 | 317 | |||
FQ223761.1 | - | 3 | 274 | |||
FQ224710.1 | - | 3 | 274 | |||
FQ224919.1 | - | 3 | 274 | |||
M20622.1 | - | 2 | 207 |
Gene Symbol: LOC100363502 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751799.3 | XP_003751847.1 | 1 | 278 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
