siRNA Details
Gene Symbol: LOC100910342 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749143.3 | XP_003749191.1 | 4 | 593 | ||
XM_006231621.2 | XP_006231683.1 | 3 | 286 | |||
XM_008760553.1 | XP_008758775.1 | 5 | 776 | |||
XM_008760554.1 | XP_008758776.1 | 5 | 750 | |||
GenBank mRNA | FQ215762.1 | - | 5 | 455 |
