siRNA Details
Gene Symbol: LOC100910342 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749143.3 | XP_003749191.1 | 4 | 581 | ||
XM_006231621.2 | XP_006231683.1 | 3 | 274 | |||
XM_008760553.1 | XP_008758775.1 | 5 | 764 | |||
XM_008760554.1 | XP_008758776.1 | 5 | 738 | |||
GenBank mRNA | FQ215762.1 | - | 5 | 443 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
