siRNA Details
Gene Symbol: Polr3gl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109571.1 | NP_001103041.1 | 6 | 544 | ||
XM_008761331.1 | XP_008759553.1 | 6 | 627 | |||
GenBank mRNA | BC168229.1 | - | 6 | 503 | 7377062 | |
FQ214367.1 | - | 6 | 530 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
