siRNA Details
Gene Symbol: LOC100366216 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750725.3 | XP_003750773.1 | 6 | 1858 | ||
XM_006245340.2 | XP_006245402.1 | 6 | 3489 | |||
XM_006245341.1 | XP_006245403.1 | 6 | 1758 | |||
XM_006245342.2 | XP_006245404.1 | 7 | 2519 | |||
XM_006245343.2 | XP_006245405.1 | 5 | 2830 | |||
XM_006245344.2 | XP_006245406.1 | 5 | 2060 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
