siRNA Details
Gene Symbol: Muc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760048.1 | XP_008758270.1 | 18 | 2946 | ||
GenBank mRNA | U07615.1 | - | 22 | 2840 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Muc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760048.1 | XP_008758270.1 | 18 | 2946 | ||
GenBank mRNA | U07615.1 | - | 22 | 2840 |