siRNA Details
Gene Symbol: Spink13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109539.1 | NP_001103009.1 | 2, 3 | 265 | ||
XM_008772175.1 | XP_008770397.1 | 2, 3 | 187 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Spink13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109539.1 | NP_001103009.1 | 2, 3 | 265 | ||
XM_008772175.1 | XP_008770397.1 | 2, 3 | 187 |