siRNA Details
Gene Symbol: Piezo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006254839.2 | XP_006254901.1 | 39 | 6459 | ||
XM_225880.8 | XP_225880.5 | 37 | 6247 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Piezo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006254839.2 | XP_006254901.1 | 39 | 6459 | ||
XM_225880.8 | XP_225880.5 | 37 | 6247 |