siRNA Details
Gene Symbol: Rcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128189.2 | NP_001121661.1 | 7 | 721 | ||
XM_006239073.2 | XP_006239135.1 | 7 | 691 | |||
XM_006239074.1 | XP_006239136.1 | 7 | 1015 | |||
XM_006239075.2 | XP_006239137.1 | 7 | 1205 | |||
XM_006239077.2 | XP_006239139.1 | 6 | 651 | |||
XM_008764172.1 | XP_008762394.1 | 7 | 2359 | |||
XM_008764173.1 | XP_008762395.1 | 8 | 745 | |||
GenBank mRNA | BC160834.1 | - | 7 | 721 | 7456254 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
