siRNA Details
Gene Symbol: Treml2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001059150.5 | XP_001059150.1 | 2 | 1021 | ||
XM_006244493.2 | XP_006244555.1 | 2 | 1022 | |||
XM_008766895.1 | XP_008765117.1 | 3 | 772 |
