siRNA Details
Gene Symbol: LOC102551340 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006227824.2 | XP_006227886.2 | 2 | 180 | ||
XM_008758708.1 | XP_008756930.1 | 2 | 181 |
Gene Symbol: Zfp748 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008758702.1 | XP_008756924.1 | 3 | 199 |

More Information
Other Species Targeted

