siRNA Details
Gene Symbol: Rnf11l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271224.1 | NP_001258153.1 | 3 | 801 |
Gene Symbol: Rnf11l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006245310.2 | XP_006245372.1 | 1 | 628 |

More Information
Other Species Targeted

Gene Ontology Categories:
