siRNA Details
Gene Symbol: Pja1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101006.1 | NP_001094476.1 | 3 | 911 | ||
XM_006257105.2 | XP_006257167.1 | 2 | 2025 | |||
XM_006257106.2 | XP_006257168.1 | 2 | 1742 | |||
XM_006257107.2 | XP_006257169.1 | 3 | 1485 | |||
GenBank mRNA | BC081885.1 | - | 3 | 911 | 7108525 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
