siRNA Details
Gene Symbol: LOC685976 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766820.1 | XP_008765042.1 | 1 | 872 | ||
XM_008766821.1 | XP_008765043.1 | 1 | 872 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
