siRNA Details
Gene Symbol: Zfp966 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762405.1 | XP_008760627.1 | 6 | 848 | ||
GenBank mRNA | AY539896.1 | - | 8 | 1067 |

More Information
Other Species Targeted

Gene Ontology Categories:
