siRNA Details
Gene Symbol: LOC103693626 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008769050.1 | XP_008767272.1 | 5 | 431 |
Gene Symbol: LOC688858 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008769051.1 | XP_008767273.1 | 566 |
