siRNA Details
Gene Symbol: Wars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168641.1 | NP_001162112.1 | 5, 6 | 641 | ||
XM_006233028.2 | XP_006233090.1 | 4, 5 | 2504 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Wars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168641.1 | NP_001162112.1 | 5, 6 | 641 | ||
XM_006233028.2 | XP_006233090.1 | 4, 5 | 2504 |