siRNA Details
Gene Symbol: Cnpy3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134710.1 | NP_001128182.1 | 3 | 311 | ||
GenBank mRNA | BC166762.1 | - | 3 | 311 | 7931606 |

More Information
Other Species Targeted

Gene Ontology Categories:
