siRNA Details
Gene Symbol: SLC25A44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286184.1 | NP_001273113.1 | 2 | 905 | ||
NM_014655.3 | NP_055470.1 | 2 | 905 | |||
XM_006711657.3 | XP_006711720.1 | 3 | 1287 | |||
XM_011510180.1 | XP_011508482.1 | 4 | 1638 | |||
XM_011510181.1 | XP_011508483.1 | 3 | 1463 | |||
XM_017002904.1 | XP_016858393.1 | 4 | 1638 | |||
XM_017002905.1 | XP_016858394.1 | 3 | 1463 | |||
XM_017002906.1 | XP_016858395.1 | 3 | 1284 | |||
XM_017002907.1 | XP_016858396.1 | 2 | 753 | |||
XM_017002908.1 | XP_016858397.1 | 3 | 1463 | |||
GenBank mRNA | AB007915.3 | - | 2 | 708 | ||
AK074912.1 | - | 2 | 750 | |||
AK075002.1 | - | 2 | 725 | |||
AK294524.1 | - | 3 | 652 | |||
AK299747.1 | - | 3 | 407 | |||
AK307968.1 | - | 4 | 517 | |||
BC008843.2 | - | 2 | 729 | 4136523 | ||
BC039854.1 | - | 2 | 708 | 6061399 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
