siRNA Details
Gene Symbol: Fam120a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191816.1 | NP_001178745.1 | 19 | 3882 |
Gene Symbol: LOC683420 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008758873.1 | XP_008757095.1 | 4 | 2380 |
