siRNA Details
Gene Symbol: LOC683422 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006252191.2 | XP_006252253.1 | 3 | 869 | ||
XM_008770796.1 | XP_008769018.1 | 3 | 283 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC683422 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006252191.2 | XP_006252253.1 | 3 | 869 | ||
XM_008770796.1 | XP_008769018.1 | 3 | 283 |