siRNA Details
Gene Symbol: LOC683469 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115040.1 | NP_001108512.1 | 3 | 572 | ||
GenBank mRNA | BC158599.1 | - | 3 | 572 | 7129113 |
Gene Symbol: LOC100909897 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771176.1 | XP_008769398.1 | 572 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
