siRNA Details
Gene Symbol: Hspb11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001131002.1 | NP_001124474.1 | 4, 5 | 394 | ||
XM_006238524.2 | XP_006238586.2 | 4, 5 | 400 | |||
XM_006238525.1 | XP_006238587.1 | 6, 7 | 659 | |||
XM_006238526.2 | XP_006238588.1 | 6, 7 | 628 | |||
XM_006238527.2 | XP_006238589.1 | 5, 6 | 561 | |||
XM_006238530.2 | XP_006238592.1 | 4, 5 | 378 | |||
XM_008763898.1 | XP_008762120.1 | 6, 7 | 645 | |||
XM_008763899.1 | XP_008762121.1 | 3, 4 | 317 | |||
XM_008763900.1 | XP_008762122.1 | 5, 6 | 544 | |||
GenBank mRNA | BC162011.1 | - | 4, 5 | 394 | 9091347 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
