siRNA Details
Gene Symbol: Hspb11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001131002.1 | NP_001124474.1 | 5 | 443 | ||
XM_006238524.2 | XP_006238586.2 | 5 | 449 | |||
XM_006238525.1 | XP_006238587.1 | 7 | 708 | |||
XM_006238526.2 | XP_006238588.1 | 7 | 677 | |||
XM_006238527.2 | XP_006238589.1 | 6 | 610 | |||
XM_006238530.2 | XP_006238592.1 | 5 | 427 | |||
XM_008763898.1 | XP_008762120.1 | 7 | 694 | |||
XM_008763899.1 | XP_008762121.1 | 4 | 366 | |||
XM_008763900.1 | XP_008762122.1 | 6 | 593 | |||
GenBank mRNA | BC162011.1 | - | 5 | 443 | 9091347 |

More Information
Other Species Targeted

Gene Ontology Categories:
