siRNA Details
Gene Symbol: Tmem25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109528.1 | NP_001102998.1 | 7, 8 | 1071 | ||
XM_006242959.2 | XP_006243021.1 | 7, 8 | 1035 | |||
XM_006242960.2 | XP_006243022.1 | 7, 8 | 1034 | |||
XM_006242961.2 | XP_006243023.1 | 7, 8 | 983 | |||
XM_008766176.1 | XP_008764398.1 | 6, 7 | 1233 |
