siRNA Details
Gene Symbol: Pisd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001061408.5 | XP_001061408.2 | 4 | 545 | ||
XM_006251423.2 | XP_006251485.1 | 4 | 4475 | |||
XM_006251424.2 | XP_006251486.1 | 4 | 3657 | |||
XM_006251426.2 | XP_006251488.1 | 3 | 4239 | |||
XM_008770389.1 | XP_008768611.1 | 3 | 3784 |

More Information
Other Species Targeted

Gene Ontology Categories:
