siRNA Details
Gene Symbol: Robo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768533.1 | XP_008766755.1 | 22 | 4263 | ||
XM_008768534.1 | XP_008766756.1 | 21 | 4236 | |||
XM_008768535.1 | XP_008766757.1 | 22 | 4098 | |||
XM_008768536.1 | XP_008766758.1 | 21 | 4071 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_019413, Y17793, CK781477.
Gene Ontology Categories:
