siRNA Details
Gene Symbol: LOC100909688 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760827.1 | XP_008759049.1 | 4 | 1426 | ||
XM_008760828.1 | XP_008759050.1 | 3 | 1330 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100909688 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760827.1 | XP_008759049.1 | 4 | 1426 | ||
XM_008760828.1 | XP_008759050.1 | 3 | 1330 |