siRNA Details
Gene Symbol: VGLL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284390.1 | NP_001271319.1 | 2 | 441 | ||
NM_014667.3 | NP_055482.2 | 2 | 441 | |||
XM_011534267.2 | XP_011532569.1 | 2 | 95 | |||
XM_017007541.1 | XP_016863030.1 | 2 | 106 | |||
XM_017007542.1 | XP_016863031.1 | 2 | 388 | |||
GenBank mRNA | AK130542.1 | - | 2 | 236 | ||
BC001514.2 | - | 2 | 206 | 2823240 | ||
BC003038.1 | - | 2 | 404 | 2822757 | ||
D50911.3 | - | 2 | 189 | |||
DA321008.1 | - | 2 | 419 | |||
DA383197.1 | - | 2 | 236 | |||
HY020255.1 | - | 2 | 441 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
