siRNA Details
Gene Symbol: VGLL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128219.2 | NP_001121691.1 | 2 | 695 | ||
NM_001284390.1 | NP_001271319.1 | 4 | 739 | |||
NM_001284391.1 | NP_001271320.1 | 2 | 412 | |||
NM_014667.3 | NP_055482.2 | 3 | 606 | |||
XM_011534267.2 | XP_011532569.1 | 4 | 381 | |||
XM_011534268.1 | XP_011532570.1 | 2 | 709 | |||
XM_011534269.1 | XP_011532571.1 | 2 | 403 | |||
XM_017007541.1 | XP_016863030.1 | 4 | 404 | |||
XM_017007542.1 | XP_016863031.1 | 3 | 553 | |||
GenBank mRNA | AK130542.1 | - | 3 | 401 | ||
AK300344.1 | - | 2 | 412 | |||
BC001514.2 | - | 3 | 371 | 2823240 | ||
BC003038.1 | - | 4 | 702 | 2822757 | ||
D50911.3 | - | 3 | 354 | |||
DA301780.1 | - | 2 | 472 | |||
DA383197.1 | - | 3 | 401 | |||
DR159250.1 | - | 2 | 329 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
