siRNA Details
Gene Symbol: Ube2e2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108371.1 | NP_001101841.1 | 3 | 336 | ||
XM_006251699.2 | XP_006251761.2 | 4 | 1141 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
